ContactUsLinkComponent
Phosphate-buffered saline (PBS) 1X
PBS Buffered Saline 1X w/o Ca & Mg, Cytiva
Catalog # 75832-012
Supplier:  Cytiva
CAS Number:  
undefined
PBS Buffered Saline 1X w/o Ca & Mg, Cytiva
Catalog # 75832-012
Supplier:  Cytiva
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    Nalgene bottle
  • Size:
    1x6 L
  • Supplier No.:
    BSS-PBS-1X6
Specifications
About this item
Cytiva manufactures PBS utilizing USP raw materials formulated in USP Water for Injection quality water.
  • GMP manufactured
  • PH: 7.1 -7.3 (USP <791>)
  • Osmolality: 280 -320 mOsm/kg (USP <785>)
Phosphate-buffered saline (PBS) 1X is a physiological salt solution commonly found in many cell culture applications. PBS is isotonic and nontoxic to cells. Cytiva offers PBS that is sterile with endotoxin levels tested for each lot.
Frequently Bought Together