ContactUsLinkComponent
TUBE CLAMP MIN 6300 PSI PVDF 1/8-3/8 IN
Pure-Fit® TC Standard Size Tube Clamps, Saint-Gobain Performance Plastics
Catalog # 75831-056
TUBE CLAMP MIN 6300 PSI PVDF 1/8-3/8 IN
Pure-Fit® TC Standard Size Tube Clamps, Saint-Gobain Performance Plastics
Catalog # 75831-056
Specifications
  • Tubing ext.Ø:
    ⅛ - ⅜"
  • Description:
    Pure-Fit® TC Tube Clamp, Assembled
  • Material:
    PVDF
  • Cat. no.:
    75831-056
  • Supplier No.:
    PFTC375PVDF
Specifications
About this item
Pure-Fit® TC tube clamps are ideal for use in the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; and a side release mechanism that facilitates secure, single-handed installation and operation.
  • Integral bore hole for lock-out capabilities
  • Provides complete fluid stoppage
  • Internal ratchet mechanism meters fluid flow
  • Can be installed on or over tubing or finished assemblies
  • Can be utilized in the flow position
  • High degree of leverage accommodates tubing with different durometers
  • Fully autoclavable and sterilizable
Constructed from high performance thermoplastic materials (PVDF or polypropylene) and available in two standard sizes to accommodate a wide range of tubing products.
Areas of use include biopharmaceutical manufacturing, pharmaceutical processes, peristaltic pump sets, drug delivery and discovery, medical systems, laboratory functions and assemblies, and tubing sets.
Certifications: Meets all USP Class VI criteria.
Customers Who Bought This Also Bought