Pure-Fit® TC Standard Size Tube Clamps, Saint-Gobain Performance Plastics
Catalog # 75831-056
Supplier: Saint Gobain Life Sciences
Specifications
- Tubing ext.Ø:⅛ - ⅜"
- Description:Pure-Fit® TC Tube Clamp, Assembled
- Material:PVDF
- Cat. no.:75831-056
- Supplier No.:PFTC375PVDF
Specifications
About this item
Pure-Fit® TC tube clamps are ideal for use in the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; and a side release mechanism that facilitates secure, single-handed installation and operation.
- Integral bore hole for lock-out capabilities
- Provides complete fluid stoppage
- Internal ratchet mechanism meters fluid flow
- Can be installed on or over tubing or finished assemblies
- Can be utilized in the flow position
- High degree of leverage accommodates tubing with different durometers
- Fully autoclavable and sterilizable
Constructed from high performance thermoplastic materials (PVDF or polypropylene) and available in two standard sizes to accommodate a wide range of tubing products.
Areas of use include biopharmaceutical manufacturing, pharmaceutical processes, peristaltic pump sets, drug delivery and discovery, medical systems, laboratory functions and assemblies, and tubing sets.
Certifications: Meets all USP Class VI criteria.
Customers Who Bought This Also Bought