Preassembled Closure and Septa GPI 13-425 Black
# de catalogue 75821-002
Fournisseur: Chemglass
Customers Who Bought This Also Bought
tert-Butyl methyl ether ≥99.5%, OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
1 062,87 $ to 3 591,58 $
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
558,88 $ to 2 230,71 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
1 313,27 $ to 5 149,78 $