Chem Snap Caps™, Colour-Coded, Chemglass
# de catalogue 75820-774
Fournisseur: Chemglass
Spécifications
- Color:Clear
- Description:Snap Cap, 10 mil PTFE Septum
- Cat. no.:75820-774
- Supplier No.:CV-3550-0011
Spécifications
A propos de cet article
These 11mm Snap Caps™ can be snapped on and off by hand, eliminating the need for metal crimping and decapping tools, and reducing potential hazards due to sharp aluminum edges.
- Large opening provides greater target area for needle penetration
- Color coded for easy sample identification
These liner-less Chem Snap Caps™ are available with or without starburst. The starburst allows for easier needle penetration. These caps were developed to handle the most volatile sample providing a liquid airtight seal to the vial. By snapping on the cap, an evaporation-proof seal of a crimp cap is achieved.
Customers Who Bought This Also Bought