Aluminum Seal Silver 20mm
# de catalogue 75820-730
Fournisseur: Chemglass
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
214,30 $ to 2 349,04 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
131,08 $ to 405,43 $
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Fournisseur: Argos Technologies
62,31 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $