Order Entry
Canada
ContactUsLinkComponent
DNA Methylation ELISA Kit, Cayman Chemical Company
Catalogue # 75817-294
Numéro CAS:  
undefined
DNA Methylation ELISA Kit, Cayman Chemical Company
Catalogue # 75817-294
Supplier Number:  589324-96
Numéro CAS:  

Spécifications

  • Assay Type:
    Competitive
  • Developmental time:
    90 - 120 min
  • Mid point:
    ~8 - 15 ng/ml (50% B/B0)
  • Detection range:
    1.17 - 150 ng/ml
  • Storage temperature:
    –20 °C
  • Sensitivity:
    ~3 ng/ml (80% B/B0)
  • Incubation time:
    18 hr
  • Cat. no.:
    75817-294
  • Supplier No.:
    589324-96
  • No. of Tests:
    96 wells

Spécifications

À propos de cet article

A competitive assay that can be used for the quantification of 5-methyl-2'-deoxy cytidine in urine, culture supernatants, plasma, and other sample matrices.

  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 5-methyl-2'-deoxy cytidine levels down to three ng/ml
  • Read: Colorimetric at 405 to 420 nm

DNA methylation is an important epigenetic process regulating gene expression. Methylation occurs on carbon 5 of 2'-deoxy-cytidine yielding the modified base 5-methyl-2'-deoxy cytidine. The methylation pattern of cells is tightly regulated during development with the methylation profile being transmitted from parent to daughter cells during cell division.

Methylation results in long-term silencing of genes, while unmethylated regions of DNA can be actively transcribed. Global changes in methylation can be quantified by measuring plasma or urinary levels of 5-methyl-2'-deoxy cytidine. These changes in methylation can provide valuable information about cancer status of an individual. For example, patients with leukemia excrete significantly elevated levels of 5-methyl-2'-deoxy cytidine compared to healthy individuals.

Global methylation within tissues can be measured in a similar manner, allowing study of tissue-specific changes that occur as a result of differentiation, aging, or carcinogenesis. Cayman’s DNA Methylation ELISA Kit is a competitive assay that can be used for the quantification of 5-methyl-2'-deoxy cytidine in urine, culture supernatants, plasma, and other sample matrices.

Delivery information: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.
Frequently Bought Together