ContactUsLinkComponent
MLA 5-D-TIPPER Pippette
MLA D-Tipper Pipettors, VistaLab
# de catalogue 75814-390
Fournisseur:  VistaLab
MLA 5-D-TIPPER Pippette
MLA D-Tipper Pipettors, VistaLab
# de catalogue 75814-390
Fournisseur:  VistaLab
Spécifications
  • Volume:
    10 µl, 50 µl, 100 µl, 200 µl, 1000 µl
  • Description:
    MLA D-Tipper Pipettor, Core Group
  • Tip Style:
    Various
  • Accuracy:
    Various
  • Precision:
    Various
  • Color Code:
    Assorted
  • Supplier No.:
    1050-0999
Spécifications
A propos de cet article
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
  • Glove guard
  • Available in a wide range of fixed volumes
  • Light plunger forces for easy operation with thumb/forefinger
  • Built-in tip ejector
  • Individually serialized
  • Lifetime warranty on parts
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
MLA pipettes are made of precision-engineered aluminum alloy and stainless steel.
These pipettors are available in 30 fixed volumes ranging from 5 to 1000 μl, or as a selectable model that is compatible at volumes of 50, 100 and 200 μl. Pipettors feature color-coded bodies for easy volume identification.
They are designed for highly accurate delivery and proven performance.
BodyTextAccessories: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Customers Who Bought This Also Bought