MLA D-Tipper Pipettors, VistaLab
# de catalogue 75814-390
Fournisseur: VistaLab
Spécifications
- Volume:10 µl, 50 µl, 100 µl, 200 µl, 1000 µl
- Description:MLA D-Tipper Pipettor, Core Group
- Tip Style:Various
- Accuracy:Various
- Precision:Various
- Color Code:Assorted
- Supplier No.:1050-0999
Spécifications
A propos de cet article
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
- Glove guard
- Available in a wide range of fixed volumes
- Light plunger forces for easy operation with thumb/forefinger
- Built-in tip ejector
- Individually serialized
- Lifetime warranty on parts
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
MLA pipettes are made of precision-engineered aluminum alloy and stainless steel.
These pipettors are available in 30 fixed volumes ranging from 5 to 1000 μl, or as a selectable model that is compatible at volumes of 50, 100 and 200 μl. Pipettors feature color-coded bodies for easy volume identification.
They are designed for highly accurate delivery and proven performance.
BodyTextAccessories: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Customers Who Bought This Also Bought