Order Entry
ContactUsLinkComponent
Lauryl Tryptose Broth with MUG (Durham Tube), 20x125mm Tube, 10ml
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
# de catalogue 75813-960
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Lauryl Tryptose Broth with MUG (Durham Tube), 20x125mm Tube, 10ml
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
# de catalogue 75813-960
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Format:
    20×125 mm tube
  • Description:
    Lauryl Tryptose w/MUG
  • Size:
    Pack of 20
  • Minimum Order Quantity:
    60
  • Supplier No.:
    K238

Spécifications

A propos de cet article

Recommended as a test of Escherichia coli and coliforms by fermentation with gas production and fluorogenic means

  • Fluorogenic test means
  • Test of Escherichia coli and coliforms
  • Gas production and MUG reaction detection

For the detection of E. coli by gas production and MUG reaction.

Customers Who Bought This Also Bought