General Frosted and Plain Microscope Slides, Epredia™
# de catalogue 89137-981
Fournisseur: Erie Scientific
Spécifications
- Description:Plain slides
- L×W:75×25 mm
- Thickness:1.0 mm
- Cat. no.:89137-981
- Supplier No.:500-001T
Spécifications
A propos de cet article
Ideal for use in nonsterile cytology and pathology applications
- Manufactured to the highest standards
- Pre-cleaned for use in hematology, microbiology, urinalysis, and general laboratory applications
- Finely ground edges
- Frosted tab versions for manual labeling or application of printed labels
- Plain versions are completely plain without frosted tab
- Ready to use
Made of water-white, noncorrosive glass for maximum clarity.
Ordering information: Packaged ½ gross per pack (two packs per gross).
Customers Who Bought This Also Bought