ContactUsLinkComponent
Screw Cap Natural for Vial 9-425 Thread
Screw Caps, 9 mm, Soft-Guard™, MicroSolv
# de catalogue 75812-820
Screw Cap Natural for Vial 9-425 Thread
Screw Caps, 9 mm, Soft-Guard™, MicroSolv
# de catalogue 75812-820
Spécifications
  • Cap Size:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), fitted
  • Material:
    PP
  • Cat. no.:
    75812-820
  • Cap Color:
    Natural
  • Supplier No.:
    9502S-E-3XN
Spécifications
A propos de cet article
Screw caps for 12 x 32 autosampler vials.
Customers Who Bought This Also Bought