Steel Insect Pins
# de catalogue 470222-076
Fournisseur: United Scientific Supplies
Spécifications
- Description:Insect pins
- Size:#0
- Cat. no.:470222-076
- Supplier No.:IPINS0-PK100
Spécifications
A propos de cet article
These steel pins are coated with a corrosion-resistant finish and have a yellow epoxy head.
- Corrosion-resistant finish
- Sold as packs of 100
- Yellow epoxy head
These steel pins are coated with a corrosion-resistant finish and have a yellow epoxy head. Sold as packs of 100.
Customers Who Bought This Also Bought