Spécifications
- Description:Goat Anti RB 800
- Cat. no.:75804-656
- Supplier No.:AC2175
Spécifications
A propos de cet article
Demo kits contain everything you need to perform single-color fluorescent Westerns
- Choose from a range of fluorescently conjugated secondary antibodies.
Kit contains 1 PVDF membrane, 10x wash buffer, 1x blocking buffer, secondary antibody and 2 background quenching sheets.
Use your own samples and primary antibodies.