ContactUsLinkComponent
Demo Kit AzureSpectra Goat-anti-rabbit 800
Azure Fluorescent Western Demo Kits, Azure Biosystems
# de catalogue 75804-656
Fournisseur:  Azure Biosystems
Product available on GSA Advantage®
Demo Kit AzureSpectra Goat-anti-rabbit 800
Azure Fluorescent Western Demo Kits, Azure Biosystems
# de catalogue 75804-656
Fournisseur:  Azure Biosystems
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Goat Anti RB 800
  • Cat. no.:
    75804-656
  • Supplier No.:
    AC2175
Spécifications
A propos de cet article
Demo kits contain everything you need to perform single-color fluorescent Westerns
  • Choose from a range of fluorescently conjugated secondary antibodies.
Kit contains 1 PVDF membrane, 10x wash buffer, 1x blocking buffer, secondary antibody and 2 background quenching sheets.
Use your own samples and primary antibodies.
Ordering information: Kit includes membrane, wash and block buffer, secondary ab, quenching sheets.
Customers Who Bought This Also Bought