ContactUsLinkComponent
Round Petri plate
Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Catalog # 75801-452
Supplier:  Corning
Round Petri plate
Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Catalog # 75801-452
Supplier:  Corning
Specifications
  • Ø:
    55 mm
  • Sterility:
    Sterile
  • Description:
    Gosselin™ Round Petri Dish with 6 Vents
  • Cat. no.:
    75801-452
  • Packaging:
    Double Outer Bag (traceable)
  • Supplier No.:
    BP53-03
Specifications
About this item
The Corning® Gosselin™ Round Petri Dish is manufactured from crystal polystyrene, and designed for use in microbiology testing applications
  • Crystal polystyrene construction
  • Double outerbag packaging
  • Easy opening sleeves
  • Sterilized to SAL 10¯6
  • Optimal flatness
The dish is designed with optimal flatness and features six vents. Manufactured in a class 100 environment.
Certifications: Food contact : Raw material is compliant with regulation EC N°1935/2004.
Sterility Assurance Level based on ISO 11137.
Customers Who Bought This Also Bought