Spécifications
- Packaging:Cubitainer
- Description:PBS 10X
- Size:20 L
- Cat. no.:75801-002
- Supplier No.:MRGF-6235-020Q
Spécifications
A propos de cet article
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 10X concentration to suit a variety of working dilution for any biological need.