ContactUsLinkComponent
PBS (Phosphate Buffered Saline) pH 7.4 10x2l
PBS (Phosphate Buffered Saline), 10X, pH 7.4, GrowCells
# de catalogue 75800-996
Fournisseur:  GROWCELLS
Numéro CAS:  
PBS (Phosphate Buffered Saline), 10X, pH 7.4, GrowCells
# de catalogue 75800-996
Fournisseur:  GROWCELLS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Polycarbonate
  • Description:
    PBS 10X
  • Size:
    2 L
  • Cat. no.:
    75800-996
  • Supplier No.:
    MRGF-6236
Spécifications
A propos de cet article
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 10X concentration to suit a variety of working dilution for any biological need.
Customers Who Bought This Also Bought