ContactUsLinkComponent
TBE (Tris-Borate EDTA)
10X TBE (Tris-Borate EDTA) pH 8.3
# de catalogue 75800-950
Fournisseur:  GROWCELLS
Numéro CAS:  
10X TBE (Tris-Borate EDTA) pH 8.3
# de catalogue 75800-950
Fournisseur:  GROWCELLS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Polycarbonate
  • Description:
    TBE 10X
  • Size:
    10 L
  • Cat. no.:
    75800-950
  • Supplier No.:
    MRGF4216010L
Spécifications
A propos de cet article
TBE is the most commonly used buffer for DNA and RNA electrophoresis
Since borate is a strong inhibitor of enzymes, TBE is recommended for DNA/RNA sizing, not for recovery of DNA for enzymatic applications. Routinely used for DNA automated sequencing gels. Used both as a running buffer and gel preparation buffer. Recommended for use with fragments smaller than 1500 bp.
Customers Who Bought This Also Bought