Spécifications
- Packaging:Polycarbonate
- Description:TBE 10X
- Size:10 L
- Cat. no.:75800-950
- Supplier No.:MRGF4216010L
Spécifications
A propos de cet article
TBE is the most commonly used buffer for DNA and RNA electrophoresis
Since borate is a strong inhibitor of enzymes, TBE is recommended for DNA/RNA sizing, not for recovery of DNA for enzymatic applications. Routinely used for DNA automated sequencing gels. Used both as a running buffer and gel preparation buffer. Recommended for use with fragments smaller than 1500 bp.