ContactUsLinkComponent
Cell Scraper 18cm
Biox Cell Scrapers and Lifters
# de catalogue 75799-934
Fournisseur:  TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Cell Scraper 18cm
Biox Cell Scrapers and Lifters
# de catalogue 75799-934
Fournisseur:  TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Description:
    Cell Scraper
  • Handle Length:
    18 cm
  • Supplier No.:
    70-1180
Spécifications
A propos de cet article
Cell Scrapers feature a uniquely designed handle and blade which reduces cell damage during the harvesting process.
  • Non-pyrogenic
  • Polyethylene construction
  • Pivoting blade makes precise cutting and scraping movements
  • Disposable
  • Pre-sterilized and individually wrapped
Our scrapers have pivot blades for precise cutting and scraping and our lifters are one sturdy piece for improved control.
Customers Who Bought This Also Bought