Biox Inoculating Loops and Needles
Loop, 10 µL, Orange
Catalog # 75799-954
Supplier: TWD Scientific
Fournisseur #: BX65-0010
Jump to:
Product Details & Documents
Innoculating loops and needles are smooth and flexible for uniform and smooth streaking, without damaging the gel surface.
- Sterile
- Polystyrene
- Other end of the loops can be used as a needle
- Free from lubricants, oils, and electrostatic charges
Needles are suitable for removal of single colony specimens and the opposite end of each loop can be used as a needle.
Spécifications
- Color:Orange
- Désignation:Loop, 10 µL