ContactUsLinkComponent
Petri Dish 90 x 15mm
Biox Petri Dishes
# de catalogue 75799-946
Fournisseur:  TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Petri Dish 90 x 15mm
Biox Petri Dishes
# de catalogue 75799-946
Fournisseur:  TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Ø:
    90 mm
  • Height:
    15 mm
  • Supplier No.:
    66-1501
Spécifications
A propos de cet article
Petri Dishes are manufactured from crystal polystyrene for optimal analysis by microscopy.
  • Easy stacking
  • Triple vented lid for improved gas exchange
  • Heavy design for optimal heat resistance
  • Clear glass-like clarity
  • Ethylene oxide sterilized
Highly transparent for microscopic analysis and vented lids for better gas exchange.
Customers Who Bought This Also Bought