Biox Cell Scrapers and Lifters
Catalogue # 75799-938
Fournisseur: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Cell Lifter
- Cat. no.:75799-938
- Handle Length:18 cm
- Supplier No.:70-2180
Spécifications
À propos de cet article
Cell Scrapers feature a uniquely designed handle and blade which reduces cell damage during the harvesting process.
- Non-pyrogenic
- Polyethylene construction
- Pivoting blade makes precise cutting and scraping movements
- Disposable
- Pre-sterilized and individually wrapped
Our scrapers have pivot blades for precise cutting and scraping and our lifters are one sturdy piece for improved control.
Frequently Bought Together