ContactUsLinkComponent
Biox Cell Scrapers and Lifters
Catalogue # 75799-938
Fournisseur:  TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Biox Cell Scrapers and Lifters
Catalogue # 75799-938
Fournisseur:  TWD Scientific
Supplier Number:  BX70-2180
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Description:
    Cell Lifter
  • Cat. no.:
    75799-938
  • Handle Length:
    18 cm
  • Supplier No.:
    70-2180
Spécifications
À propos de cet article
Cell Scrapers feature a uniquely designed handle and blade which reduces cell damage during the harvesting process.
  • Non-pyrogenic
  • Polyethylene construction
  • Pivoting blade makes precise cutting and scraping movements
  • Disposable
  • Pre-sterilized and individually wrapped
Our scrapers have pivot blades for precise cutting and scraping and our lifters are one sturdy piece for improved control.
Frequently Bought Together