VWR® Premium Plain and Frosted Micro Slides
Catalog # 75799-268
Supplier: VWR International
Specifications
- Description:Plain slides, economy
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Cat. no.:75799-268
Specifications
About this item
These pre-cleaned slides are made from the highest-quality sheet glass.
- Standard and Economy formats available
- Glass fabrication
Economy slides are made from a lesser-quality sheet glass for greater cost-effectiveness.
Customers Who Bought This Also Bought