Prevail* Rocket 53x7mm
# de catalogue 75798-948
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
109,33 $ to 364,87 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
46,59 $ to 465,92 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
377,44 $ to 992,90 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
194,14 $ to 694,12 $
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
120,60 $ to 125,07 $