Vydac 214TP C4, HPLC Columns
Catalog # 75798-756
Supplier: HICHROM LIMITED
Specifications
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Pore Size:300 Å
- Length:250 mm
- USP code:L26
- Column Hardware:Stainless Steel
- Cat. no.:75798-756
- Packing Material:Silica
- Phase:214TP C4
- Supplier No.:214TP54
Specifications
About this item
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L26
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
Frequently Bought Together