Alltima* HP C8 Rocket 53x7mm
# de catalogue 75797-672
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
63,72 $ to 462,44 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
131,08 $ to 405,43 $
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
131,41 $ to 136,31 $
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
279,61 $
VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening
Fournisseur: VWR International
73,97 $ to 705,67 $