VWR® Micro Cover Glasses, Round, No. 2
# de catalogue 48382-042
Fournisseur: VWR International
Spécifications
- Ø:18 mm
- Description:Cover glasses, round
- Cat. no.:48382-042
Spécifications
A propos de cet article
These microscope cover glasses are corrosion-resistant and uniformly thin and flat.
- Borosilicate glass fabrication
- Thickness: 0.17-0.25mm
The processing techniques result in superior cleanliness. Manufactured from borosilicate glass, cover glasses are free from bubbles, scratches, and striations.
Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought