VWR® Premium Superfrost® Colorfrost® Microscope Slides
Catalog # 48312-401
Supplier: VWR International
Specifications
- Color:Blue
- Description:Colorfrost slides
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Cat. no.:48312-401
Specifications
About this item
VWR® Superfrost® and Colorfrost® Micro Slides feature brilliant coloring and a coating that accepts and retains laboratory markings better than comparable products.
- Easy to write on, easy to read
- Key shows which side is up
- Eliminate sticking and scratching
Slides are frosted and have a brilliant colored, opaque coating
All slides have the word “SPECIMEN” printed on them, except for the white Premium Superfrost slide, which is unprinted.
All slides are 75x25mm and are autoclavable.
Coating takes and retains most laboratory marking methods better than plain or conventional frosted slides. The coating is impervious to most common laboratory chemicals and reagents. Slides feature a raised printed writing surface that eliminates the need for paper labels and re-labeling of slides.
Ordering information: For additional sizes and thicknesses, contact your Avantor sales representative.
Customers Who Bought This Also Bought