ContactUsLinkComponent
VWR* Plain Precleaned Micro Slide
VWR® Premium Plain and Frosted Micro Slides
Catalog # 48300-031
Supplier:  VWR International
undefined
VWR® Premium Plain and Frosted Micro Slides
Catalog # 48300-031
Supplier:  VWR International
Specifications
  • Description:
    Plain slides
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.2 mm
  • Cat. no.:
    48300-031
Specifications
About this item
These pre-cleaned slides are made from the highest-quality sheet glass.
  • Standard and Economy formats available
  • Glass fabrication
Economy slides are made from a lesser-quality sheet glass for greater cost-effectiveness.
Frequently Bought Together