VWR® Premium Plain and Frosted Micro Slides
Catalogue # 48300-026
Fournisseur: VWR International
Spécifications
- Description:Plain slides
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Cat. no.:48300-026
Spécifications
À propos de cet article
These pre-cleaned slides are made from the highest-quality sheet glass.
- Standard and Economy formats available
- Glass fabrication
Economy slides are made from a lesser-quality sheet glass for greater cost-effectiveness.
Frequently Bought Together