N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH83636.400
Fournisseur: VWR International
Numéro CAS: 127-19-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, poly-coated
- Size:4 L
- Cat. no.:BDH83636.400
Spécifications
Résultats des tests de spécification
- Assay (by GC-FID):Min 99.5%
- Residue after evaporation:Max 5 ppm
- Water (by KF coulometry):Max 0.03%
- Transmittance at 270 nm:Min 10 T(%)
- Transmittance at 280 nm:Min 50 T(%)
- Transmittance at 290 nm:Min 70 T(%)
- Transmittance at 310 nm:Min 90 T(%)
- Transmittance at 320 nm:Min 93 T(%)
- Transmittance at 360 nm:Min 97 T(%)
- Transmittance at 400 nm:Min 97 T(%)
- Optical absorbance at 270 nm:Max 1.00 AU
- Optical absorbance at 280 nm:Max 0.30 AU
- Optical absorbance at 290 nm:Max 0.15 AU
- Optical absorbance at 320 AU:Max 0.05 AU
- Optical absorbance at 310 nm:Max 0.03 AU
- Optical absorbance at 360 nm:Max 0.01 AU
- Optical absorbance at 400 nm:Max 0.01 AU
- N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Acetic acid dimethylamide
Point d'ébullition:166,1 °C (1013 hPa)
Point de Fusion:-20 °C
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
Point d'éclair:70 °C
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 166,1 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -20 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 70 °C
:M.W. 87,12 g/mol
:N,N-Diméthylacétamide
:-20
:166,1
Point d'ébullition:166.1 °C (1013 hPa)
Point de Fusion:–20 °C
Point d'éclair:70 °C
ONU:2810
:70
:99,5
:Boiling Pt:
:Boiling Pt: 166.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –20 °C
:Flash Pt:
:Flash Pt: 70 °C
:M.W. 87.12 g/mol
:N,N-Dimethylacetamide
:166.1
:6.1
:III
Customers Who Bought This Also Bought