Lysing Matrix E, MP Biomedicals
Catalogue # 75784-630
Fournisseur: MP Biomedicals
Spécifications
- Description:Lysing Matrix E
- Size:100×2ml
- Adapter compatibility:QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Instrument Compatibility:FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- Tube compatibility:2 ml
- Key applications:Extractions of nucleic acids, proteins, metabolites and other small molecules from soil, fecal, wastwater, sludge and other environmental samples
- Application areas:ample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Material of construction:1.4 ceramic spheres, 0.1 mm silica spheres, and 4 mm glass beads.
- Matrix type:E
- Molecule enriched:DNA; RNA; Proteins; Other small molecules
- Materials required (not supplied):FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Storage and handling:Store at Room Temperature (15...30 °C)
- Sample volume:up to 500 mg per prep
- Sample type:Soil; Feces; Environmental Water; Wastewater; Sludge
- Specific gravity:Ceramic Spheres, 4; Silica Spheres, 2.5; Glass Bead, 2.5
- Product type:Lab Essentials
- Cat. no.:75784-630
- Supplier No.:116914100
Spécifications
À propos de cet article
Lysing Matrix E is effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
- Impact resistant tube
- Contains 1.4mm ceramic spheres, 0.1mm silica spheres and 4mm glass beads
- DNase and RNase Free
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.
Frequently Bought Together