To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Lysing Matrix D
- Size:50×2 ml
- Adapter compatibility:QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Instrument Compatibility:FastPrep-24; FastPrep 120; FastPrep-96
- Tube compatibility:2 ml
- Key applications:Lysis of soft tissues
- Application areas:Nucleic Acid Isolation & Purification
- Material of construction:Matrix D, contains 1.4 mm ceramic spheres.
- Matrix type:D
- Molecule enriched:DNA; RNA; Proteins; Other small molecules
- Materials required (not supplied):FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Storage and handling:Store at Room Temperature (15...30 °C)
- Sample type:Plants; Animal Tissues; Cultured Cells
- Specific gravity:4
- Dimensions:1.2 - 1.6 mm
- Product type:Lab Essentials
- Cat. no.:75784-626
- Supplier No.:116913050
Spécifications
A propos de cet article
Lysing matrix D is for rapid and reliable biological sample lysis from a wide variety of starting materials.
- Impact resistant tube containing 1.4mm ceramic spheres
- Primarily used for the lysis of softer tissues
- Will also lyse plant tissue, cultured cells and whole insects
- Grinding matrix is chemically inert and will not bind nucleic acids
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.