ContactUsLinkComponent
PAD FOAM BIOPSY POLYESTER BL 30.2X25X2MM
Blue Biopsy Pads, Jaece
# de catalogue 75783-972
Fournisseur:  Jaece
PAD FOAM BIOPSY POLYESTER BL 30.2X25X2MM
Blue Biopsy Pads, Jaece
# de catalogue 75783-972
Fournisseur:  Jaece
Spécifications
  • Dimensions:
    30.2 × 25 × 2 mm
  • Color:
    Blue
  • Description:
    Biopsy Pad
  • Cat. no.:
    75783-972
  • Supplier No.:
    BP-1001
Spécifications
A propos de cet article
Blue Biopsy Pads allow fluids to transfer safely and thoroughly around the tissue while holding the specimen secure.
  • Made from uniform polyester reticulated open cell foam
  • Transfer fluids safely around the tissue
  • Reduced risk of tissue sticking during embedding
  • Verified for consistency
Blue Biopsy Pads are made from uniform polyester reticulated open cell foam, which allows fluids to transfer safely and thoroughly around the tissue while holding the specimen secure.
The thin 2 mm design helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding.
Customers Who Bought This Also Bought