Xylene (mixture of isomers), low water content 98.5-99.41% (isomers plus ethylbenzene) ACS, VWR Chemicals BDH®
Catalog # BDH1170-4LG
Supplier: VWR International
CAS Number: 1330-20-7
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:BDH1170-4LG
- Supplier No.:PL371ZA-4
Specifications
Specification Test Results
- GC Purity (Xylenes + Ethylbenzene:98.5% Min. 99.41 %
- Ethylbenzene (by GC):Less than 25.0%
- Color (APHA):Less than 10
- Residue:Less than 0.002%
- Sub. Darkened by H2SO4:Passes Test
- Sulfur Compounds (as S:Less than 0.003%
- Water (by K.F.):Less than 0.02%
- VWR Chemicals BDH®, Xylene (mixture of isomers), low water content 98.5-99.41% (isomers plus ethylbenzene) ACS:
Specification Test Results
About this item
Boiling Pt:140 °C (1013 hPa)
Melting Pt:-34 - 34 °C
Density:0,86 to 0.88 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
Flash Pt:25 °C
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 140 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -34 à 34 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 25 °C
:M.W. 106,17 g/mol
:140
UN:1307
:25
:12,10214
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86 to 0.88 g/cm³ (20 °C)
:98,5-99,41
Flash Pt:25 °C
:Boiling Pt:
:Boiling Pt: 140 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –34 °C
:Flash Pt:
:Flash Pt: 25 °C
:M.W. 106.17 g/mol
:3
:III
Customers Who Bought This Also Bought