ContactUsLinkComponent
Neoprene Bunsen Burner Hose/Tubing 3 ft
Neoprene Burner Tubing
# de catalogue 470220-806
Fournisseur:  Wards
Neoprene Bunsen Burner Hose/Tubing 3 ft
Neoprene Burner Tubing
# de catalogue 470220-806
Fournisseur:  Wards
Spécifications
  • Description:
    Neoprene connecting hose, 90 cm
  • Cat. no.:
    470220-806
  • Supplier No.:
    CH0100B
Spécifications
A propos de cet article
Connect your burner to a fuel source easily and safely.
  • Two sizes available
  • Tight secure connection to most common burners and gas sources
This neoprene hose connects your bunsen, tirrill or any other type of burner to a fuel source.
Customers Who Bought This Also Bought