ContactUsLinkComponent
Petri plate square 4 vent
Corning® Gosselin™ Square Petri Dish, 120×120 mm, Corning
# de catalogue 10799-777
Fournisseur:  Corning
Petri plate square 4 vent
Corning® Gosselin™ Square Petri Dish, 120×120 mm, Corning
# de catalogue 10799-777
Fournisseur:  Corning
Spécifications
  • Dimensions:
    120 x 120 mm
  • Description:
    Gosselin™ Square Petri Dish
  • Cat. no.:
    10799-777
  • Packaging:
    14/Sleeve, 252/Case
  • Supplier No.:
    BP124-05
Spécifications
A propos de cet article
PS. Petri plates manufactured under aseptic cleanroom conditions.
  • Manufactured from optically clear virgin polystyrene
  • Sterilized by irradiation to SAL 10⁻³
  • Supplied with 4 vents to provide consistent gas exchange
  • New design to enhance stacked dish stability
Corning® Gosselin™ Square petri dishes are optically clear, with a design that has been optimized for use on automated platforms.
The square design is used in colony picking and antibiogram applications.
Customers Who Bought This Also Bought