Flexible Feel Inoculation Loops in Tamper-Evident Resealable Bags, Globe Scientific
# de catalogue 10799-759
Fournisseur: Globe Scientific
Spécifications
- Capacity:10 µl
- Sterility:Sterile
- Cat. no.:10799-759
- Packaging:10/Zip Lock Bag, 100 Bags/Case
- Supplier No.:2875-10
Spécifications
A propos de cet article
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media.
- Gamma sterilized, sterility assurance level 10-3
- Certificate of calibration
- Tamper-evident resealable bags
- Each bag individually labeled with lot number and expiration date
- Integrated loop/needle design
The loops are packaged in tamper-evident resealable bags. Each bag is individually labeled with lot number and expiration date. All loops are sterilized by gamma irradiation. The 1 µl and 10 µl inoculation loops are available with the loop on one end and an integrated needle on the other end, eliminating the need for a separate needle.
The polystyrene (PS) inoculation loops are smooth and flexible to facilitate uniform streaking of culture media.
Certifications: Manufactured under ISO 9001:2008 and ISO 14001:2004 standards
Customers Who Bought This Also Bought