ContactUsLinkComponent
Inoculation Loop
Flexible Feel Inoculation Loops in Tamper-Evident Resealable Bags, Globe Scientific
# de catalogue 10799-759
Fournisseur:  Globe Scientific
Inoculation Loop
Flexible Feel Inoculation Loops in Tamper-Evident Resealable Bags, Globe Scientific
# de catalogue 10799-759
Fournisseur:  Globe Scientific
Spécifications
  • Capacity:
    10 µl
  • Sterility:
    Sterile
  • Cat. no.:
    10799-759
  • Packaging:
    10/Zip Lock Bag, 100 Bags/Case
  • Supplier No.:
    2875-10
Spécifications
A propos de cet article
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media.
  • Gamma sterilized, sterility assurance level 10-3
  • Certificate of calibration
  • Tamper-evident resealable bags
  • Each bag individually labeled with lot number and expiration date
  • Integrated loop/needle design
The loops are packaged in tamper-evident resealable bags. Each bag is individually labeled with lot number and expiration date. All loops are sterilized by gamma irradiation. The 1 µl and 10 µl inoculation loops are available with the loop on one end and an integrated needle on the other end, eliminating the need for a separate needle.
The polystyrene (PS) inoculation loops are smooth and flexible to facilitate uniform streaking of culture media.
Certifications: Manufactured under ISO 9001:2008 and ISO 14001:2004 standards
Customers Who Bought This Also Bought