Multi-Layer Foil Gas Sampling Bags, Restek
Catalogue # 10863-020
Fournisseur: Restek
Spécifications
- Capacity:5 L
- Description:Gas sampling bag, multi-layer foil, with polypropylene combo valve and septum
- Size:12×12"
- Cat. no.:10863-020
- Supplier No.:22952
Spécifications
À propos de cet article
Sampling bags are a low-cost, whole-air sampling device for high-level VOCs and permanent gases.
- Good stability for low molecular weight compounds
- Chemically inert with light and moisture protection
- Not recommended for low ppm VOCs due to background levels
- Protective five layer barrier minimizes gas permeability
- 60 gauge nylon (outer layer)
- Metalized aluminum/Polyethylene
- 0.0003" aluminum foil/0.002" polyethylene (inner layer)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for your application. All our bags feature a polypropylene combo valve with hose connection to fit 3/16" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Frequently Bought Together