ContactUsLinkComponent
HazMat Mat Pad
HazMat Mat Pad, PIG®
Catalog # 10860-084
Supplier:  New Pig
HazMat Mat Pad
HazMat Mat Pad, PIG®
Catalog # 10860-084
Supplier:  New Pig
Specifications
  • Description:
    PIG® HazMat Mat Pad in Bench Box® Dispenser
  • Cat. no.:
    10860-084
  • Absorbency:
    12.8 oz./pad
  • Supplier No.:
    MAT3510
Specifications
About this item
The HazMat Mat Pad absorbs the widest range of acids, bases, corrosives, chemicals and unknown liquids.
  • Chemical-resistant mat won't degrade
  • Won't rip, tear or fray even when saturated
  • Absorbs high concentrations like 98% sulfuric acid and 30% sodium hydroxide
Absorbs the widest range of acids, bases, corrosives, chemicals and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated.
Customers Who Bought This Also Bought