Spécifications
- Size:950 ml (32 oz.)
- Packaging:Poly Bottle
- Supplier No.:4102
Spécifications
A propos de cet article
Conflikt ® Ready-to-use Disinfectant Spray cleans and disinfects hard surfaces that may harbor viruses, bacteria and fungi
Pre-diluted, stable solution of dual quaternary ammonium active ingredients is effective against HIV-1, E-coli and other pathogenic organisms. Conforms to OSHA's Bloodborne Pathogens Standard. Conflikt is effective against a wide range of organisms including TB. EPA registered.