ContactUsLinkComponent
Conflikt* Ready-To-Use Dosinfectant 950 mL
Conflikt, Ready-to-use Disinfectant, Decon Labs
# de catalogue CA89233-180
Fournisseur:  Decon Labs
Conflikt* Ready-To-Use Dosinfectant 950 mL
Conflikt, Ready-to-use Disinfectant, Decon Labs
# de catalogue CA89233-180
Fournisseur:  Decon Labs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    950 ml (32 oz.)
  • Packaging:
    Poly Bottle
  • Supplier No.:
    4102
Spécifications
A propos de cet article
Conflikt ® Ready-to-use Disinfectant Spray cleans and disinfects hard surfaces that may harbor viruses, bacteria and fungi
Pre-diluted, stable solution of dual quaternary ammonium active ingredients is effective against HIV-1, E-coli and other pathogenic organisms. Conforms to OSHA's Bloodborne Pathogens Standard. Conflikt is effective against a wide range of organisms including TB. EPA registered.
Customers Who Bought This Also Bought