High Visibility Absorbent Mat Pad, PIG®
# de catalogue 10860-054
Fournisseur: New Pig
Spécifications
- Description:PIG® High-visibility absorbent mat pad
- Cat. no.:10860-054
- Absorbency:22 gal./bag
- Dimensions:15" Wx20" L
- Supplier No.:MAT603
Spécifications
A propos de cet article
Hi-viz spill absorbents alert workers to potential hazards or repairs.
- High-visibility yellow color
- Won't rip, tear or fray even when saturated
- Absorbs most common industrial liquids
Hi-viz spill absorbents alert workers to potential hazards or repairs. The high-visibility yellow color is easier to see in poorly lit areas to minimize tripping hazards; alerts workers to potential dangers and work in progress. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind. Absorbs most common industrial liquids - oils, water, solvents, coolants and more.
Customers Who Bought This Also Bought