Universal Cylinder Wrench, Restek
# de catalogue 10855-804
Fournisseur: Restek
Spécifications
- Description:Universal Cylinder Wrench
- Cat. no.:10855-804
- Supplier No.:21322
Spécifications
A propos de cet article
Use this versatile wrench for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.
- Multiple sizes allow for universal matching with any system
Customers Who Bought This Also Bought