Inserts for Versa Vials™, Restek
# de catalogue 10854-366
Fournisseur: Avantor
Spécifications
- Cap included:No
- Capacity:0.25 ml
- Description:Versa vial insert with flange
- Material:PP
- Bottom Style:Conical bottom
- Supplier No.:22710
Spécifications
A propos de cet article
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
- 250 µl volume vial compatibility
- Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
Customers Who Bought This Also Bought