11 mm Poly Crimp Seal Caps, Snap-On or Crimp, Restek
# de catalogue 10854-300
Fournisseur: Restek
Spécifications
- Color:Clear
- Description:Snap Top Vial Caps
- Cat. no.:10854-300
- Supplier No.:26569
- Orifice:11 mm
Spécifications
A propos de cet article
Large, economical packs of Restek Poly Crimp seal caps are available for easy application onto any crimp-top vial.
- Snap-on or crimp-top versatility
- Preslit caps available to reduce risk of needle bending
- Clear, blue, or mixed colors available
The Poly Crimp seal is versatile, working as either a snap-on or crimp-top cap. Simply use a standard crimping tool to secure the Poly Crimp seal onto any crimp-top vial.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought