ContactUsLinkComponent
Crimper Jaw Set, 11mm, For use With High Power Crimping Tool
Accessories for High-Power Electronic Crimping Tool, Restek
# de catalogue 10854-198
Fournisseur:  Restek
Accessories for High-Power Electronic Crimping Tool, Restek
# de catalogue 10854-198
Fournisseur:  Restek
Spécifications
  • Description:
    Interchangeable Crimper Jaw Set for High-Powered Electronic Crimping Tool
  • Cat. no.:
    10854-198
  • Seal Size:
    11 mm
  • Supplier No.:
    23424