ContactUsLinkComponent
Magnetic Caps and Septa for SPME 18 mm
Magnetic Screw-Thread Caps, 18 mm, Restek
# de catalogue 10854-176
Fournisseur:  Restek
Magnetic Caps and Septa for SPME 18 mm
Magnetic Screw-Thread Caps, 18 mm, Restek
# de catalogue 10854-176
Fournisseur:  Restek
Spécifications
  • Description:
    Magnetic Caps and Septa for SPME
  • Cat. no.:
    10854-176
  • Supplier No.:
    23091
  • Orifice:
    18 mm
Spécifications
A propos de cet article
18 mm magnetic screw-thread caps are quickly applied by magnetic autosamplers and feature wide orifices for ensuring efficient needle insertion without breaking.
  • Economical
  • Sturdy fabrication
  • Wide orifice for easy needle insertion
These screw-thread caps are compatible with magnetic autosamplers, making application easier for the user.
Customers Who Bought This Also Bought