Order Entry
ContactUsLinkComponent
Polypropylene Vial Storage Rack 36 vial capacity
Vial Rack for Headspace Vials, Restek
# de catalogue 10854-124
Fournisseur:  Restek
Polypropylene Vial Storage Rack 36 vial capacity
Vial Rack for Headspace Vials, Restek
# de catalogue 10854-124
Fournisseur:  Restek

Spécifications

  • Capacity:
    36 vial capacity
  • Description:
    Polypropylene Vial Storage Rack for 6, 10, or 20 mL Headspace Vials
  • Cat. no.:
    10854-124
  • Supplier No.:
    22644

Spécifications

A propos de cet article

These vial racks securely holds up to 36 headspace vials.

  • Autoclavable up to 121°C for 20 minutes
  • Vials lock in rack for convenient one-handed cap removal
  • Easy to clean and fully autoclavable
  • Stackable even with vials in place

The openings in the bottom of each well facilitate drainage and the alphanumeric indexing on rack simplifies sample identification.

Customers Who Bought This Also Bought