Order Entry
ContactUsLinkComponent
WISP 48 Seal with Septa 13 mm
Aluminum Seals with Septa for 4.0 ml WISP Crimp-Top Vials, Restek
# de catalogue 10854-074
Fournisseur:  Restek
WISP 48 Seal with Septa 13 mm
Aluminum Seals with Septa for 4.0 ml WISP Crimp-Top Vials, Restek
# de catalogue 10854-074
Fournisseur:  Restek

Spécifications

  • Color:
    Silver
  • Description:
    WISP 48 Seals w/Septa
  • Material:
    Aluminum
  • Cat. no.:
    10854-074
  • Supplier No.:
    21755
  • Orifice:
    13 mm

Spécifications

A propos de cet article

Aluminum seals provide a sturdy cap to crimp-top vials, with wide orifices for easy needle insertion.

  • Crimp-top vial compatibility
  • Economical
  • Silver aluminum

Aluminum seals are the preferred choice for use with manual crimping tools, and for autosamplers that do not employ magnetic pick-up transport.

Wide 13 mm openings decrease the number of broken needles, and aluminum fabrication imbues these seals with great durability.

Customers Who Bought This Also Bought