Order Entry
ContactUsLinkComponent
WISP 48 Cap and Septa, Preassembled, Type: Screw-Thread, Septa Material: Red PTFE/Silicone
WISP 48 Caps and Septa, Restek
# de catalogue 10854-062
Fournisseur:  Restek
WISP 48 Cap and Septa, Preassembled, Type: Screw-Thread, Septa Material: Red PTFE/Silicone
WISP 48 Caps and Septa, Restek
# de catalogue 10854-062
Fournisseur:  Restek

Spécifications

  • Description:
    WISP 48 Caps and Septa, Top Hat
  • Cat. no.:
    10854-062
  • Supplier No.:
    21745
  • Orifice:
    13 mm/425

Spécifications

A propos de cet article

These replacement caps and septa are compatible with WISP vials (largely screw-thread) and serve as economical alternatives to purchasing new whole units.

  • Economical
  • Sturdy construction
  • Wide orifices for easier needle insertion

Durable, functional features make these inexpensive replacement parts an indispensable addition to any set of WISP vials.

Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought