WISP 48 Caps and Septa, Restek
# de catalogue 10854-058
Fournisseur: Restek
Spécifications
- Description:WISP 48 Caps
- Material:Black Polypropylene, Open-Hole
- Cat. no.:10854-058
- Supplier No.:21742
- Orifice:13 mm/425
Spécifications
A propos de cet article
These replacement caps and septa are compatible with WISP vials (largely screw-thread) and serve as economical alternatives to purchasing new whole units.
- Economical
- Sturdy construction
- Wide orifices for easier needle insertion
Durable, functional features make these inexpensive replacement parts an indispensable addition to any set of WISP vials.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought