VWR® Tapes, PP
# de catalogue 10853-868
Fournisseur: VWR International
Spécifications
- Length:36 yds.
- Width:2"
- Color:Clear
- Cat. no.:10853-868
Spécifications
A propos de cet article
VWR® polypropylene tapes are designed with acrylic adhesive.
- Wound on plastic core
Customers Who Bought This Also Bought